Product Details

SNP ID
rs113235029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51078960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCGGGGTACCTGGCTGGGGGA[A/C]ACTGCAGGGTTATAACTGGGTCTAC
Phenotype
MIM: 606135
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLK14 PubMed Links

Gene Details

Gene
KLK14
Gene Name
kallikrein related peptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311182.1 Intron NP_001298111.1
NM_022046.5 Intron NP_071329.2
XM_017026820.1 Intron XP_016882309.1

View Full Product Details