Product Details

SNP ID
rs113032403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51993444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCAGTGAAGCCTTTTCCACA[C/G]TCATTGCAAATATAAGGTTTCTCTC
Phenotype
MIM: 605422
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF350 PubMed Links

Gene Details

Gene
ZNF350
Gene Name
zinc finger protein 350
There are no transcripts associated with this gene.

Gene
ZNF615
Gene Name
zinc finger protein 615
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199324.1 1982 Missense Mutation GAC,GAG D555E NP_001186253.1
NM_001321317.1 1982 Missense Mutation GAC,GAG D549E NP_001308246.1
NM_001321318.1 1982 Missense Mutation GAC,GAG D551E NP_001308247.1
NM_001321319.1 1982 Missense Mutation GAC,GAG D555E NP_001308248.1
NM_001321320.1 1982 Missense Mutation GAC,GAG D544E NP_001308249.1
NM_001321321.1 1982 Missense Mutation GAC,GAG D555E NP_001308250.1
NM_001321322.1 1982 Missense Mutation GAC,GAG D551E NP_001308251.1
NM_001321323.1 1982 Missense Mutation GAC,GAG D560E NP_001308252.1
NM_198480.3 1982 Missense Mutation GAC,GAG D544E NP_940882.3
XM_011526824.1 1982 Missense Mutation GAC,GAG D555E XP_011525126.1
XM_011526825.1 1982 Missense Mutation GAC,GAG D555E XP_011525127.1
XM_011526826.1 1982 Missense Mutation GAC,GAG D555E XP_011525128.1
XM_011526829.1 1982 Missense Mutation GAC,GAG D436E XP_011525131.1
XM_011526830.1 1982 Missense Mutation GAC,GAG D436E XP_011525132.1
XM_017026648.1 1982 Missense Mutation GAC,GAG D544E XP_016882137.1
XM_017026649.1 1982 Missense Mutation GAC,GAG D544E XP_016882138.1
XM_017026650.1 1982 Missense Mutation GAC,GAG D436E XP_016882139.1
XM_017026651.1 1982 Missense Mutation GAC,GAG D436E XP_016882140.1

View Full Product Details