Product Details

SNP ID
rs115728501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52398506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGAAGTGGGCATCTTATTCCAAT[C/T]CCCTCCCTGTGAATGTGTGGAGAAA
Phenotype
MIM: 615580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF528 PubMed Links

Gene Details

Gene
ZNF528
Gene Name
zinc finger protein 528
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032423.2 169 UTR 5 NP_115799.2
XM_005259323.1 169 Intron XP_005259380.1
XM_006723418.2 169 UTR 5 XP_006723481.1
XM_011527392.1 169 UTR 5 XP_011525694.1
XM_011527394.2 169 Intron XP_011525696.1
XM_017027360.1 169 UTR 5 XP_016882849.1
XM_017027361.1 169 UTR 5 XP_016882850.1
XM_017027362.1 169 UTR 5 XP_016882851.1
XM_017027363.1 169 UTR 5 XP_016882852.1
XM_017027364.1 169 UTR 5 XP_016882853.1
XM_017027365.1 169 Intron XP_016882854.1
Gene
ZNF528-AS1
Gene Name
ZNF528 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF880
Gene Name
zinc finger protein 880
There are no transcripts associated with this gene.

View Full Product Details