Product Details
- SNP ID
-
rs114959714
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57377307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGAGCATGACGGAACACACCCC[A/G]AGCAGGGACTGTACACGTGTCCAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF547
PubMed Links
Gene Details
- Gene
- ZNF547
- Gene Name
- zinc finger protein 547
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173631.3 |
612 |
Missense Mutation |
AAG,GAG |
K111E |
NP_775902.2 |
View Full Product Details