Product Details

SNP ID
rs117167880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4658139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACATTGAAAATTCTCAGAAGTC[C/T]GGAGGATTTGGGGTGGGGCCCATGG
Phenotype
MIM: 606746 MIM: 615869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYDGF PubMed Links

Gene Details

Gene
MYDGF
Gene Name
myeloid derived growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019107.3 Intron NP_061980.1
XM_017026987.1 Intron XP_016882476.1
Gene
TNFAIP8L1
Gene Name
TNF alpha induced protein 8 like 1
There are no transcripts associated with this gene.

View Full Product Details