Product Details

SNP ID
rs117030331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5565594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCTCATAGAAGGTACGCTGCC[A/G]AGAACACCCTCTCCTAGCCCCAAGC
Phenotype
MIM: 615241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TINCR PubMed Links

Gene Details

Gene
TINCR
Gene Name
tissue differentiation-inducing non-protein coding RNA
There are no transcripts associated with this gene.

View Full Product Details