Product Details

SNP ID
rs113689392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6374826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGCAGGGGCTCAGCCCGTTA[C/T]GACTTCTCCCATGAGATCCTTCGGG
Phenotype
MIM: 613305 MIM: 601119 MIM: 189968 MIM: 602921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH7 PubMed Links

Gene Details

Gene
ALKBH7
Gene Name
alkB homolog 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032306.3 483 Silent Mutation TAC,TAT Y173Y NP_115682.1
XM_005259658.4 483 Silent Mutation TAC,TAT Y147Y XP_005259715.1
XM_017027355.1 483 Missense Mutation ACG,ATG T247M XP_016882844.1
Gene
CLPP
Gene Name
caseinolytic mitochondrial matrix peptidase proteolytic subunit
There are no transcripts associated with this gene.

Gene
GTF2F1
Gene Name
general transcription factor IIF subunit 1
There are no transcripts associated with this gene.

Gene
PSPN
Gene Name
persephin
There are no transcripts associated with this gene.

View Full Product Details