Product Details

SNP ID
rs115300336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31848507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGGTTGAGGACCCATCTGGG[C/T]GCTCGGTTGAGGCACTTAGAGTGCA
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 968 Intron NP_001012662.1
NM_001320478.1 968 Intron NP_001307407.1
NM_001320479.1 968 Missense Mutation CAC,CGC H262R NP_001307408.1
NM_080611.4 968 Intron NP_542178.2
NM_177991.2 968 Intron NP_817130.1
XM_017027654.1 968 Missense Mutation CAC,CGC H302R XP_016883143.1
XM_017027655.1 968 Missense Mutation CAC,CGC H299R XP_016883144.1
XM_017027656.1 968 Intron XP_016883145.1
XM_017027657.1 968 Missense Mutation CAC,CGC H265R XP_016883146.1
XM_017027658.1 968 Missense Mutation CAC,CGC H199R XP_016883147.1
XM_017027659.1 968 Missense Mutation CAC,CGC H199R XP_016883148.1
XM_017027660.1 968 Missense Mutation CAC,CGC H199R XP_016883149.1
XM_017027661.1 968 Missense Mutation CAC,CGC H199R XP_016883150.1
XM_017027662.1 968 Missense Mutation CAC,CGC H199R XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
There are no transcripts associated with this gene.

View Full Product Details