Product Details

SNP ID
rs115623645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:299805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAAAGAGAAGGTTTTTAAGGA[A/G]GGGCTTCTGAATACTTGGGAGATAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
There are no transcripts associated with this gene.

Gene
ZCCHC3
Gene Name
zinc finger CCHC-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033089.6 2243 UTR 3 NP_149080.2

View Full Product Details