Product Details

SNP ID
rs117023834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3108579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTGCATCAAGGTCAGTGCCAA[A/G]AGGAAGCAGAGCAAAGCATGTGACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBOX5 PubMed Links

Gene Details

Gene
UBOX5
Gene Name
U-box domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267584.1 3177 UTR 3 NP_001254513.1
NM_014948.3 3177 UTR 3 NP_055763.1
NM_199415.2 3177 UTR 3 NP_955447.1
Gene
UBOX5-AS1
Gene Name
UBOX5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details