Product Details

SNP ID
rs113229055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38741053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAAAGCTCTCCACTCCATCAG[A/G]CCTCTAATTCCACACCTGGAAAAAA
Phenotype
MIM: 611723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00114 PubMed Links

Gene Details

Gene
LINC00114
Gene Name
long intergenic non-protein coding RNA 114
There are no transcripts associated with this gene.

View Full Product Details