Product Details

SNP ID
rs115095004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42168712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCGCGGTCTTGTTCTGCAAGG[A/G]GGGGAGAGGGCACGGAAGGGGAGGC
Phenotype
MIM: 603107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCF20 PubMed Links

Gene Details

Gene
TCF20
Gene Name
transcription factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005650.3 6145 Missense Mutation ACC,GCC T1942A NP_005641.1
NM_181492.2 6145 Intron NP_852469.1
XM_005261722.3 6145 Missense Mutation ACC,GCC T1942A XP_005261779.1
XM_006724313.3 6145 Missense Mutation ACC,GCC T1942A XP_006724376.1
XM_011530353.2 6145 Intron XP_011528655.1
XM_011530354.2 6145 Intron XP_011528656.1

View Full Product Details