Product Details

SNP ID
rs113924126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43133254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCTGCTCCCACTTCACTGGGGAG[A/G]CCAGCTGCTGGGCCAGCAGCTTGTG
Phenotype
MIM: 603392 MIM: 614479
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIK PubMed Links

Gene Details

Gene
BIK
Gene Name
BCL2 interacting killer
There are no transcripts associated with this gene.

Gene
MCAT
Gene Name
malonyl-CoA-acyl carrier protein transacylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014507.3 1011 UTR 3 NP_055322.1
NM_173467.4 1011 Missense Mutation GCC,GTC A321V NP_775738.3
XM_011530156.2 1011 Missense Mutation GCC,GTC A156V XP_011528458.1

View Full Product Details