Product Details

SNP ID
rs116160154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183649958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAACACTTGACAATTTAAACAC[G/T]TCTTTAGAAAAATCACGTGTGCCTA
Phenotype
MIM: 614138
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RWDD4 PubMed Links

Gene Details

Gene
RWDD4
Gene Name
RWD domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307922.1 Intron NP_001294851.1
NM_152682.3 Intron NP_689895.2
Gene
TRAPPC11
Gene Name
trafficking protein particle complex 11
There are no transcripts associated with this gene.

View Full Product Details