Product Details

SNP ID
rs117160246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109443674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTTCTGTACCCCTCAGTAAAA[A/G]AGGAGAGGACTGTCAGCACTGAACA
Phenotype
MIM: 607184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC24B PubMed Links

Gene Details

Gene
SEC24B
Gene Name
SEC24 homolog B, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042734.3 Intron NP_001036199.1
NM_001300813.2 Intron NP_001287742.1
NM_001318085.1 Intron NP_001305014.1
NM_001318086.1 Intron NP_001305015.1
NM_006323.4 Intron NP_006314.2
XM_005262688.1 Intron XP_005262745.1
XM_005262691.1 Intron XP_005262748.1
XM_011531535.2 Intron XP_011529837.1
XM_011531536.2 Intron XP_011529838.1
XM_011531537.2 Intron XP_011529839.1
XM_011531539.2 Intron XP_011529841.1
XM_011531540.2 Intron XP_011529842.1
XM_017007652.1 Intron XP_016863141.1
Gene
SEC24B-AS1
Gene Name
SEC24B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details