Product Details

SNP ID
rs116565777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112238237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTATCTTACATTTGTTGAATG[A/C]ACAGCCAGAAGAGTTATTTGAACTT
Phenotype
MIM: 610851
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP1AR PubMed Links

Gene Details

Gene
AP1AR
Gene Name
adaptor related protein complex 1 associated regulatory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128426.2 Intron NP_001121898.1
NM_018569.5 Intron NP_061039.3
XM_017008378.1 Intron XP_016863867.1
XM_017008379.1 Intron XP_016863868.1
XM_017008380.1 Intron XP_016863869.1
Gene
LOC105377371
Gene Name
uncharacterized LOC105377371
There are no transcripts associated with this gene.

View Full Product Details