Product Details

SNP ID
rs115964925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152537613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCATTAAAATGCTGCGGGAAAG[C/G]TGGTGAGAGGGGCAAAGGAGGATGT
Phenotype
MIM: 606278
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DKFZP434I0714 PubMed Links

Gene Details

Gene
DKFZP434I0714
Gene Name
uncharacterized protein DKFZP434I0714
There are no transcripts associated with this gene.

Gene
FBXW7
Gene Name
F-box and WD repeat domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013415.1 Intron NP_001013433.1
NM_001257069.1 Intron NP_001243998.1
NM_018315.4 Intron NP_060785.2
NM_033632.3 Intron NP_361014.1
Gene
MIR4453
Gene Name
microRNA 4453
There are no transcripts associated with this gene.

View Full Product Details