Product Details
- SNP ID
-
rs114651703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:43231286 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTGCAGAGGTATTTCACAAAGA[C/T]CAGGGTCTCGGCCCAAAAGTTGACT
- Phenotype
-
MIM: 611800
MIM: 612053
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01126
PubMed Links
Gene Details
- Gene
- LINC01126
- Gene Name
- long intergenic non-protein coding RNA 1126
There are no transcripts associated with this gene.
- Gene
- THADA
- Gene Name
- THADA, armadillo repeat containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001083953.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1842V |
NP_001077422.1 |
NM_001271643.1 |
5876 |
Intron |
|
|
NP_001258572.1 |
NM_001271644.1 |
5876 |
Intron |
|
|
NP_001258573.1 |
NM_022065.4 |
5876 |
Missense Mutation |
ATC,GTC |
I1842V |
NP_071348.3 |
XM_006712061.3 |
5876 |
Missense Mutation |
ATC,GTC |
I1842V |
XP_006712124.1 |
XM_006712062.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1841V |
XP_006712125.1 |
XM_006712063.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1802V |
XP_006712126.1 |
XM_006712064.2 |
5876 |
Missense Mutation |
ATC,GTC |
I1798V |
XP_006712127.1 |
XM_006712065.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1768V |
XP_006712128.1 |
XM_006712066.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1721V |
XP_006712129.1 |
XM_006712067.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1720V |
XP_006712130.1 |
XM_006712068.3 |
5876 |
Intron |
|
|
XP_006712131.1 |
XM_006712069.3 |
5876 |
Missense Mutation |
ATC,GTC |
I1025V |
XP_006712132.1 |
XM_011533015.2 |
5876 |
Intron |
|
|
XP_011531317.1 |
XM_017004674.1 |
5876 |
Missense Mutation |
ATC,GTC |
I1801V |
XP_016860163.1 |
XM_017004675.1 |
5876 |
Missense Mutation |
ATC,GTC |
I946V |
XP_016860164.1 |
- Gene
- ZFP36L2
- Gene Name
- ZFP36 ring finger protein like 2
There are no transcripts associated with this gene.
View Full Product Details