Product Details
- SNP ID
-
rs115529610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:73203602 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTGAACTTGAGCTACACACTTC[A/G]CTCTGAGCCTGCGTGCATGCAATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOTO
PubMed Links
Gene Details
- Gene
- NOTO
- Gene Name
- notochord homeobox
View Full Product Details