Product Details

SNP ID
rs115295641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28758586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATTTCAAGGCTTATTCAAGTAT[A/G]AATAACCAGTAGCCAGTTTTTTCCT
Phenotype
MIM: 600590
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1CB PubMed Links

Gene Details

Gene
PPP1CB
Gene Name
protein phosphatase 1 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002709.2 Intron NP_002700.1
NM_206876.1 Intron NP_996759.1

View Full Product Details