Product Details

SNP ID
rs115747406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179919180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTTGCCCATAGTGGTTCTGTA[C/T]TGGGATTAGCAGTAAGATGCAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF130 PubMed Links

Gene Details

Gene
RNF130
Gene Name
ring finger protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280801.1 2825 Intron NP_001267730.1
NM_018434.5 2825 Intron NP_060904.2
XM_011534593.2 2825 UTR 3 XP_011532895.1

View Full Product Details