Product Details

SNP ID
rs114405829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181239024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTGGCTAAGTGCTCCTTCCATG[A/G]CGCTGTCTTCCACTGAGTAGGAGAC
Phenotype
MIM: 176981 MIM: 610530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTC-338M12.4 PubMed Links

Gene Details

Gene
CTC-338M12.4
Gene Name
uncharacterized LOC101928649
There are no transcripts associated with this gene.

Gene
RACK1
Gene Name
receptor for activated C kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006098.4 Intron NP_006089.1
Gene
SNORD95
Gene Name
small nucleolar RNA, C/D box 95
There are no transcripts associated with this gene.

Gene
SNORD96A
Gene Name
small nucleolar RNA, C/D box 96A
There are no transcripts associated with this gene.

Gene
TRIM41
Gene Name
tripartite motif containing 41
There are no transcripts associated with this gene.

View Full Product Details