Product Details

SNP ID
rs111699274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:37103635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGTGGAATTCTAAAGATAATCT[G/T]GTAGTTACTAAGATTTTCTTATGTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1A1L PubMed Links

Gene Details

Gene
CSNK1A1L
Gene Name
casein kinase 1 alpha 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145203.5 2030 UTR 3 NP_660204.2

View Full Product Details