Product Details

SNP ID
rs114593377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49444740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTCTCAGGGTGCAGCCTTAAT[A/G]AGAGGTGATTCCTAAGCTGCTGGGA
Phenotype
MIM: 612175 MIM: 607865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAB39L PubMed Links

Gene Details

Gene
CAB39L
Gene Name
calcium binding protein 39 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079670.2 448 Intron NP_001073138.1
NM_001287337.1 448 Intron NP_001274266.1
NM_001287338.1 448 Intron NP_001274267.1
NM_001287339.1 448 Intron NP_001274268.1
NM_030925.3 448 Intron NP_112187.2
XM_011535254.2 448 Intron XP_011533556.1
XM_011535255.2 448 Intron XP_011533557.1
XM_011535256.2 448 Intron XP_011533558.1
XM_017020785.1 448 Intron XP_016876274.1
XM_017020786.1 448 Intron XP_016876275.1
XM_017020787.1 448 Intron XP_016876276.1
XM_017020788.1 448 Intron XP_016876277.1
Gene
SETDB2
Gene Name
SET domain bifurcated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160308.2 448 UTR 5 NP_001153780.1
NM_001320699.1 448 UTR 5 NP_001307628.1
NM_031915.2 448 UTR 5 NP_114121.2
Gene
SETDB2-PHF11
Gene Name
SETDB2-PHF11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320727.1 448 UTR 5 NP_001307656.1

View Full Product Details