Product Details

SNP ID
rs114353705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39267228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCGGGTCGGGCTCGGACCTGC[G/T]CTGCCTCGGGATGTAAAGTATAACA
Phenotype
MIM: 602132
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTAGE5 PubMed Links

Gene Details

Gene
CTAGE5
Gene Name
CTAGE family member 5, ER export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247988.1 105 Intron NP_001234917.1
NM_001247989.1 105 UTR 5 NP_001234918.1
NM_001247990.1 105 UTR 5 NP_001234919.1
NM_005930.3 105 UTR 5 NP_005921.2
NM_203354.2 105 Intron NP_976229.1
NM_203355.2 105 UTR 5 NP_976230.1
NM_203356.2 105 Intron NP_976231.1
XM_005267648.4 105 Intron XP_005267705.1
XM_006720148.2 105 Intron XP_006720211.1
XM_011536775.2 105 Intron XP_011535077.1
XM_011536778.2 105 Intron XP_011535080.1
XM_011536784.2 105 Intron XP_011535086.1
XM_011536785.2 105 Intron XP_011535087.1
XM_017021314.1 105 Intron XP_016876803.1
XM_017021315.1 105 Intron XP_016876804.1
XM_017021316.1 105 Intron XP_016876805.1
XM_017021317.1 105 Intron XP_016876806.1
XM_017021318.1 105 Intron XP_016876807.1
XM_017021319.1 105 Intron XP_016876808.1
XM_017021320.1 105 Intron XP_016876809.1
XM_017021321.1 105 Intron XP_016876810.1
XM_017021322.1 105 Intron XP_016876811.1
XM_017021323.1 105 Intron XP_016876812.1
XM_017021324.1 105 Intron XP_016876813.1
XM_017021325.1 105 Intron XP_016876814.1
XM_017021326.1 105 Intron XP_016876815.1
XM_017021327.1 105 Intron XP_016876816.1
XM_017021328.1 105 Intron XP_016876817.1
XM_017021329.1 105 Intron XP_016876818.1
XM_017021330.1 105 Intron XP_016876819.1
Gene
LOC100288846
Gene Name
uncharacterized LOC100288846
There are no transcripts associated with this gene.

View Full Product Details