Product Details

SNP ID
rs113626637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50312145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAACCAAATAACGCAGCGCTGG[A/C]ACCATCCCCTACGCACGCTCCCCTC
Phenotype
MIM: 609584
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5S PubMed Links

Gene Details

Gene
ATP5S
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003803.2 85 Intron NP_001003803.1
NM_001003805.2 85 Intron NP_001003805.1
NM_015684.3 85 Intron NP_056499.2
XM_005267537.3 85 Intron XP_005267594.1
XM_011536649.2 85 Intron XP_011534951.1
XM_011536650.2 85 Intron XP_011534952.1
XM_011536651.2 85 Intron XP_011534953.1
XM_011536652.1 85 Intron XP_011534954.1
XM_011536653.2 85 Intron XP_011534955.1
XM_011536654.2 85 Intron XP_011534956.1
XM_011536655.2 85 Intron XP_011534957.1
XM_011536656.2 85 Intron XP_011534958.1
XM_011536657.2 85 Intron XP_011534959.1
XM_017021220.1 85 Intron XP_016876709.1
Gene
L2HGDH
Gene Name
L-2-hydroxyglutarate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024884.2 85 Silent Mutation GTG,GTT V2V NP_079160.1
XM_005268075.4 85 Silent Mutation GTG,GTT V2V XP_005268132.1
XM_011537166.2 85 UTR 5 XP_011535468.1
XM_011537167.2 85 Intron XP_011535469.1
XM_011537168.2 85 Intron XP_011535470.1
XM_017021655.1 85 Intron XP_016877144.1
XM_017021656.1 85 UTR 5 XP_016877145.1
XM_017021657.1 85 UTR 5 XP_016877146.1
XM_017021658.1 85 Silent Mutation GTG,GTT V2V XP_016877147.1

View Full Product Details