Product Details

SNP ID
rs117883794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:61695560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGACAGGAGGATCACCCTCTT[A/C]GTCGCTTCGGCCAGTGTGTCGGGCT
Phenotype
MIM: 603348 MIM: 614528
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HIF1A PubMed Links

Gene Details

Gene
HIF1A
Gene Name
hypoxia inducible factor 1 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243084.1 160 Intron NP_001230013.1
NM_001530.3 160 UTR 5 NP_001521.1
NM_181054.2 160 UTR 5 NP_851397.1
Gene
HIF1A-AS1
Gene Name
HIF1A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details