Product Details

SNP ID
rs111906529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101023366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGACCGTATAGCGTACGCTTTA[C/T]CTGTGACGTATGTAACACGGTCCAC
Phenotype
MIM: 616358 MIM: 613654 MIM: 616036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1193 PubMed Links

Gene Details

Gene
MIR1193
Gene Name
microRNA 1193
There are no transcripts associated with this gene.

Gene
MIR1197
Gene Name
microRNA 1197
There are no transcripts associated with this gene.

Gene
MIR299
Gene Name
microRNA 299
There are no transcripts associated with this gene.

Gene
MIR323A
Gene Name
microRNA 323a
There are no transcripts associated with this gene.

Gene
MIR329-1
Gene Name
microRNA 329-1
There are no transcripts associated with this gene.

Gene
MIR329-2
Gene Name
microRNA 329-2
There are no transcripts associated with this gene.

Gene
MIR379
Gene Name
microRNA 379
There are no transcripts associated with this gene.

Gene
MIR380
Gene Name
microRNA 380
There are no transcripts associated with this gene.

Gene
MIR411
Gene Name
microRNA 411
There are no transcripts associated with this gene.

Gene
MIR494
Gene Name
microRNA 494
There are no transcripts associated with this gene.

Gene
MIR543
Gene Name
microRNA 543
There are no transcripts associated with this gene.

Gene
MIR758
Gene Name
microRNA 758
There are no transcripts associated with this gene.

View Full Product Details