Product Details

SNP ID
rs114901832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140180765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTGTGGGCAGCAAGCTTGTGG[C/T]ATGACTTTGCCAAAACAACAGATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYSTM1 PubMed Links

Gene Details

Gene
CYSTM1
Gene Name
cysteine rich transmembrane module containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032412.3 Intron NP_115788.1

View Full Product Details