Product Details

SNP ID
rs117989175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57621339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGGTCAAGGCGCTGACCAGCG[A/G]GGCCGCGCTGCACGTGCTCTTGTCC
Phenotype
MIM: 610215 MIM: 601873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF25 PubMed Links

Gene Details

Gene
ARHGEF25
Gene Name
Rho guanine nucleotide exchange factor 25
There are no transcripts associated with this gene.

Gene
B4GALNT1
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
There are no transcripts associated with this gene.

Gene
LOC101927583
Gene Name
uncharacterized LOC101927583
There are no transcripts associated with this gene.

Gene
SLC26A10
Gene Name
solute carrier family 26 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133489.2 778 Missense Mutation GAG,GGG E156G NP_597996.2

View Full Product Details