Product Details

SNP ID
rs111260250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69740076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTGGAGGAGGGGCCCATAAGA[A/G]GATTACAACAAAACTGTTCCATTGT
Phenotype
MIM: 608686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928002 PubMed Links

Gene Details

Gene
LOC101928002
Gene Name
uncharacterized LOC101928002
There are no transcripts associated with this gene.

Gene
RAB3IP
Gene Name
RAB3A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024647.3 Intron NP_001019818.1
NM_001278402.1 Intron NP_001265331.1
NM_022456.4 Intron NP_071901.2
NM_175623.3 Intron NP_783322.1
NM_175624.3 Intron NP_783323.1
NM_175625.3 Intron NP_783324.1
XM_006719226.3 Intron XP_006719289.3
XM_006719227.3 Intron XP_006719290.3
XM_017018773.1 Intron XP_016874262.1
XM_017018774.1 Intron XP_016874263.1
XM_017018775.1 Intron XP_016874264.1
XM_017018776.1 Intron XP_016874265.1
XM_017018777.1 Intron XP_016874266.1

View Full Product Details