Product Details

SNP ID
rs117324318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71440257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTCCGACCTGGGGCTCTCGGA[A/G]CTGCCTTCCAACCTCAGCGTCTTCA
Phenotype
MIM: 606667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGR5 PubMed Links

Gene Details

Gene
LGR5
Gene Name
leucine rich repeat containing G protein-coupled receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277226.1 488 Silent Mutation GAA,GAG E59E NP_001264155.1
NM_001277227.1 488 Silent Mutation GAA,GAG E59E NP_001264156.1
NM_003667.3 488 Silent Mutation GAA,GAG E59E NP_003658.1

View Full Product Details