Product Details

SNP ID
rs112754090
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114063562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGCAGATTCTGGAGTATGCA[C/T]ATACAGCCACGCTGCAAGCCAAGGC
Phenotype
MIM: 176797
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 966 Missense Mutation CAT,TAT H88Y NP_001018011.1
NM_006006.4 966 Missense Mutation CAT,TAT H88Y NP_005997.2
XM_005271658.4 966 Missense Mutation CAT,TAT H88Y XP_005271715.1
XM_017018257.1 966 Missense Mutation CAT,TAT H88Y XP_016873746.1
XM_017018258.1 966 Missense Mutation CAT,TAT H88Y XP_016873747.1
XM_017018259.1 966 Missense Mutation CAT,TAT H88Y XP_016873748.1

View Full Product Details