Product Details

SNP ID
rs114562244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123884104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGACAAGAAGAAAACATTTAAG[C/T]AAGCAGTATACATGATCCAGGTGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM225 PubMed Links

Gene Details

Gene
TMEM225
Gene Name
transmembrane protein 225
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013743.2 704 Missense Mutation TAC,TGC Y145C NP_001013765.2
XM_006718832.3 704 Missense Mutation TAC,TGC Y95C XP_006718895.1
XM_011542802.2 704 Intron XP_011541104.1
XM_017017653.1 704 Intron XP_016873142.1

View Full Product Details