Product Details

SNP ID
rs115536685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102713283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTATAAGCAGGACACAATGTAA[C/T]GAAAATGCTTGCTGAACTTCCCTTC
Phenotype
MIM: 120355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
There are no transcripts associated with this gene.

Gene
MMP8
Gene Name
matrix metallopeptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304441.1 1648 UTR 3 NP_001291370.1
NM_001304442.1 1648 UTR 3 NP_001291371.1
NM_002424.2 1648 UTR 3 NP_002415.1
XM_011542834.2 1648 UTR 3 XP_011541136.1
XM_011542835.2 1648 UTR 3 XP_011541137.1
XM_011542836.2 1648 UTR 3 XP_011541138.1
XM_017017771.1 1648 UTR 3 XP_016873260.1

View Full Product Details