Product Details

SNP ID
rs114440759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:110738407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTACAGCCTTAGAAAAGGAAAA[C/T]GCGTAATTTCTAACTCCAGGAGGAC
Phenotype
MIM: 610826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A46 PubMed Links

Gene Details

Gene
SLC25A46
Gene Name
solute carrier family 25 member 46
There are no transcripts associated with this gene.

Gene
TMEM232
Gene Name
transmembrane protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039763.3 Intron NP_001034852.3
XM_006714670.3 Intron XP_006714733.1
XM_011543552.2 Intron XP_011541854.1
XM_011543553.2 Intron XP_011541855.1
XM_011543555.2 Intron XP_011541857.1
XM_011543556.1 Intron XP_011541858.1
XM_011543557.2 Intron XP_011541859.1
XM_011543559.2 Intron XP_011541861.1
XM_011543560.2 Intron XP_011541862.1
XM_011543561.2 Intron XP_011541863.1
XM_011543563.2 Intron XP_011541865.1
XM_011543564.2 Intron XP_011541866.1
XM_011543565.2 Intron XP_011541867.1
XM_011543566.2 Intron XP_011541868.1
XM_011543567.2 Intron XP_011541869.1
XM_017009704.1 Intron XP_016865193.1
XM_017009705.1 Intron XP_016865194.1
XM_017009706.1 Intron XP_016865195.1
XM_017009707.1 Intron XP_016865196.1

View Full Product Details