Product Details

SNP ID
rs111331676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7670257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGCACTTGTGGCTCCGCCTGT[G/T]GCTCGTGGGATGTGCGCGCCGAGAC
Phenotype
MIM: 609565 MIM: 605565
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MCEMP1 PubMed Links

Gene Details

Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
There are no transcripts associated with this gene.

Gene
RETN
Gene Name
resistin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193374.1 272 Missense Mutation GGC,TGC G79C NP_001180303.1
NM_020415.3 272 Missense Mutation GGC,TGC G79C NP_065148.1

View Full Product Details