Product Details

SNP ID
rs114182400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:128097886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAATATAGTACAAGTCAGTAC[C/T]GCTGATTTCTGCATTCATTCAGCAA
Phenotype
MIM: 176741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MKI67 PubMed Links

Gene Details

Gene
MKI67
Gene Name
marker of proliferation Ki-67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145966.1 9171 UTR 3 NP_001139438.1
NM_002417.4 9171 UTR 3 NP_002408.3
XM_006717864.3 9171 UTR 3 XP_006717927.2
XM_011539818.2 9171 UTR 3 XP_011538120.1

View Full Product Details