Product Details

SNP ID
rs117851550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94929271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTCCAGGAACATGTGTACAG[C/T]TTCCTGCCAGGGGCTGAAGTATGGG
Phenotype
MIM: 601130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2C9 PubMed Links

Gene Details

Gene
CYP2C9
Gene Name
cytochrome P450 family 2 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000771.3 Intron NP_000762.2
XM_017015758.1 Intron XP_016871247.1

View Full Product Details