Product Details

SNP ID
rs137958414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75094793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGTCTGGATGGCCTCTACCAG[C/T]GTCATGTTCTCACAGATCATGAGGA
Phenotype
MIM: 613191 MIM: 611575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP13 PubMed Links

Gene Details

Gene
DUSP13
Gene Name
dual specificity phosphatase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007271.1 984 UTR 3 NP_001007272.1
NM_001007272.1 984 Silent Mutation NP_001007273.1
NM_001007273.1 984 Silent Mutation NP_001007274.1
NM_001320842.1 984 Silent Mutation NP_001307771.1
NM_001320843.1 984 Silent Mutation NP_001307772.1
NM_016364.3 984 Silent Mutation NP_057448.3
XM_005269883.3 984 Silent Mutation XP_005269940.1
XM_005269884.4 984 Silent Mutation XP_005269941.3
XM_005269887.1 984 Silent Mutation XP_005269944.1
XM_005269890.1 984 Silent Mutation XP_005269947.1
XM_011539853.1 984 Silent Mutation XP_011538155.1
XM_011539854.2 984 Silent Mutation XP_011538156.1
XM_011539855.1 984 Silent Mutation XP_011538157.1
XM_011539856.2 984 Silent Mutation XP_011538158.1
XM_017016313.1 984 Silent Mutation XP_016871802.1
XM_017016314.1 984 Intron XP_016871803.1
Gene
SAMD8
Gene Name
sterile alpha motif domain containing 8
There are no transcripts associated with this gene.

View Full Product Details