Product Details

SNP ID
rs139127110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118594155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGATGACCACGCTGACGGTCA[C/T]ATTCTGGCCATGGGGGGCTATCTTG
Phenotype
MIM: 600895
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRLHR PubMed Links

Gene Details

Gene
PRLHR
Gene Name
prolactin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004248.2 1229 Missense Mutation ATG,GTG M364V NP_004239.1

View Full Product Details