Product Details
- SNP ID
-
rs143033065
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:101828163 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTGCCCTGCACCTCCTTGGTGAT[A/G]CAGCCGTCCTTGTTAAGGTCATACA
- Phenotype
-
MIM: 604661
MIM: 604039
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KCNIP2
PubMed Links
Gene Details
- Gene
- KCNIP2
- Gene Name
- potassium voltage-gated channel interacting protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014591.4 |
742 |
Silent Mutation |
TGC,TGT |
C210C |
NP_055406.2 |
NM_173191.2 |
742 |
Silent Mutation |
TGC,TGT |
C195C |
NP_775283.1 |
NM_173192.2 |
742 |
Silent Mutation |
TGC,TGT |
C177C |
NP_775284.1 |
NM_173193.2 |
742 |
Silent Mutation |
TGC,TGT |
C152C |
NP_775285.1 |
NM_173194.2 |
742 |
Silent Mutation |
TGC,TGT |
C150C |
NP_775286.1 |
NM_173195.2 |
742 |
Silent Mutation |
TGC,TGT |
C145C |
NP_775287.1 |
NM_173197.2 |
742 |
Intron |
|
|
NP_775289.1 |
XM_005269729.2 |
742 |
Silent Mutation |
TGC,TGT |
C177C |
XP_005269786.1 |
XM_005269730.2 |
742 |
Silent Mutation |
TGC,TGT |
C145C |
XP_005269787.1 |
XM_006717812.2 |
742 |
Silent Mutation |
TGC,TGT |
C195C |
XP_006717875.1 |
XM_011539731.2 |
742 |
Silent Mutation |
TGC,TGT |
C150C |
XP_011538033.1 |
XM_017016161.1 |
742 |
Silent Mutation |
TGC,TGT |
C155C |
XP_016871650.1 |
- Gene
- KCNIP2-AS1
- Gene Name
- KCNIP2 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MGEA5
- Gene Name
- meningioma expressed antigen 5 (hyaluronidase)
There are no transcripts associated with this gene.
View Full Product Details