Product Details

SNP ID
rs143477537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118594627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACTCCTCGCAGAGGCGCACGTC[A/G]TGCGGCTTGAGCTCCACGTGATAGG
Phenotype
MIM: 600895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRLHR PubMed Links

Gene Details

Gene
PRLHR
Gene Name
prolactin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004248.2 757 Silent Mutation CAC,CAT H206H NP_004239.1

View Full Product Details