Product Details

SNP ID
rs146429924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122697880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTAAGTCACCAGGGAATTCTGA[C/T]ATGCAGAAAAGATTTCGGTTTGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf120 PubMed Links

Gene Details

Gene
C10orf120
Gene Name
chromosome 10 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010912.2 893 Missense Mutation ATA,ATG I287M NP_001010912.1

View Full Product Details