Product Details

SNP ID
rs146432350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45616437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACAGTAAGATGTAGAGGAAGAGT[G/T]AGATTTTTGGAAGCTTTGGTGCATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFAND4 PubMed Links

Gene Details

Gene
ZFAND4
Gene Name
zinc finger AN1-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128324.2 2435 Silent Mutation NP_001121796.1
NM_001282905.1 2435 Silent Mutation NP_001269834.1
NM_001282906.1 2435 Silent Mutation NP_001269835.1
NM_174890.3 2435 Silent Mutation NP_777550.2
XM_017016935.1 2435 Intron XP_016872424.1

View Full Product Details