Product Details

SNP ID
rs149625416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92452545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCTGGAAATTGGAAAATGCTGA[C/G]GCATACCAAACAATTGCAGGTTCAT
Phenotype
MIM: 146680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IDE PubMed Links

Gene Details

Gene
IDE
Gene Name
insulin degrading enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165946.1 3619 UTR 3 NP_001159418.1
NM_001322793.1 3619 UTR 3 NP_001309722.1
NM_001322794.1 3619 UTR 3 NP_001309723.1
NM_001322795.1 3619 UTR 3 NP_001309724.1
NM_001322796.1 3619 UTR 3 NP_001309725.1
NM_001322797.1 3619 UTR 3 NP_001309726.1
NM_004969.3 3619 UTR 3 NP_004960.2
XM_017016187.1 3619 UTR 3 XP_016871676.1
XM_017016188.1 3619 UTR 3 XP_016871677.1
XM_017016189.1 3619 UTR 3 XP_016871678.1
XM_017016190.1 3619 UTR 3 XP_016871679.1

View Full Product Details