Product Details

SNP ID
rs149880380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102511469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCTGCCTTTGCATAGCGACCA[A/G]TTCTGGAGATGATGATGGTATCATA
Phenotype
MIM: 605143 MIM: 607035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
There are no transcripts associated with this gene.

Gene
SUFU
Gene Name
SUFU negative regulator of hedgehog signaling
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178133.1 Intron NP_001171604.1
NM_016169.3 Intron NP_057253.2
XM_011539858.2 Intron XP_011538160.1
XM_011539860.2 Intron XP_011538162.1
XM_011539861.2 Intron XP_011538163.1
XM_011539863.2 Intron XP_011538165.1
XM_011539864.2 Intron XP_011538166.1
XM_017016323.1 Intron XP_016871812.1

View Full Product Details