Product Details
- SNP ID
-
rs150078715
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92463846 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCTAATGCCTGAATGTGTTTTTG[C/G]AAGGCCTCTTCTGTCATGTCCTCTA
- Phenotype
-
MIM: 146680
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IDE
PubMed Links
Gene Details
- Gene
- IDE
- Gene Name
- insulin degrading enzyme
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001165946.1 |
1306 |
Missense Mutation |
TTC,TTG |
F327L |
NP_001159418.1 |
NM_001322793.1 |
1306 |
Missense Mutation |
TTC,TTG |
F882L |
NP_001309722.1 |
NM_001322794.1 |
1306 |
Missense Mutation |
TTC,TTG |
F843L |
NP_001309723.1 |
NM_001322795.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
NP_001309724.1 |
NM_001322796.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
NP_001309725.1 |
NM_001322797.1 |
1306 |
Missense Mutation |
TTC,TTG |
F327L |
NP_001309726.1 |
NM_004969.3 |
1306 |
Missense Mutation |
TTC,TTG |
F882L |
NP_004960.2 |
XM_017016187.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
XP_016871676.1 |
XM_017016188.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
XP_016871677.1 |
XM_017016189.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
XP_016871678.1 |
XM_017016190.1 |
1306 |
Missense Mutation |
TTC,TTG |
F841L |
XP_016871679.1 |
View Full Product Details