Product Details

SNP ID
rs112110823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73269948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGGGAGCAGAGGTGGCTTTG[C/T]CTTTTCGGAAGAACTGGTTCTGTGG
Phenotype
MIM: 602451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RY6 PubMed Links

Gene Details

Gene
P2RY6
Gene Name
pyrimidinergic receptor P2Y6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277204.1 1346 Intron NP_001264133.1
NM_001277205.1 1346 Intron NP_001264134.1
NM_001277206.1 1346 Intron NP_001264135.1
NM_001277207.1 1346 Intron NP_001264136.1
NM_001277208.1 1346 Intron NP_001264137.1
NM_176796.2 1346 Intron NP_789766.1
NM_176797.2 1346 Intron NP_789767.1
NM_176798.2 1346 UTR 5 NP_789768.1
XM_005274022.3 1346 Intron XP_005274079.1
XM_006718571.3 1346 Intron XP_006718634.1
XM_011545076.2 1346 UTR 5 XP_011543378.1
XM_011545077.2 1346 UTR 5 XP_011543379.1
XM_011545079.2 1346 Intron XP_011543381.1

View Full Product Details