Product Details

SNP ID
rs112770452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93148617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGCCAGCTACAACTTTGGGAG[C/T]AGGATAAATAATTTCTTCAACAGTT
Phenotype
MIM: 613760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC36A4 PubMed Links

Gene Details

Gene
SLC36A4
Gene Name
solute carrier family 36 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286139.1 1578 Missense Mutation ACT,GCT T344A NP_001273068.1
NM_152313.3 1578 Missense Mutation ACT,GCT T479A NP_689526.2
XM_017017174.1 1578 UTR 3 XP_016872663.1
XM_017017175.1 1578 Missense Mutation ACT,GCT T373A XP_016872664.1
XM_017017176.1 1578 Missense Mutation ACT,GCT T344A XP_016872665.1
XM_017017177.1 1578 UTR 3 XP_016872666.1

View Full Product Details